Learn More
Cole-Parmer Cleanroom Male Luer Lock Plug and Nut Assemblies, Straight Adapter, White Nylon
Alle ansehen Cole-Parmer Produkte
Click to view available options
Farbe:
Beere
Gelb
Lila
Limette
Naturfarben
Orange
Rosé
Packungsgröße:
10 Stück
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Nut and body rotate independently during connection
Spezifikation
Spezifikation
| Zertifizierung/Konformität | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Material | Nylon |
| Einlass | Male Luer Lock, Plug |
| Farbe | Beere |