Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Y Union, White Nylon
Alle ansehen Cole-Parmer Produkte
Click to view available options
Innendurchmesser (angloamerikanisch):
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/8 in.
5/8 in.
Packungsgröße:
10 Stück
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. FDA and RoHS compliant
Spezifikation
Spezifikation
| Zertifizierung/Konformität | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Innendurchmesser (angloamerikanisch) | 3/8 in. |
| Verpackungsmenge | 10/Pk. |
| Auslassanschluss | Hose Barb Y |
| Sterilisationsmethode | E-beam/gamma irradiation, EtO, or autoclave up to 121°C |
| Form | Y |
| Farbe | White |