missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cirazoline hydrochloride, Tocris Bioscience™
Click to view available options
Menge:
10 mg
Beschreibung
Selective α1 agonist.
Spezifikation
Spezifikation
Chemischer Name oder Material | Cirazoline hydrochloride |
CAS | 40600-13-3 |
Summenformel | C13H17ClN2O |
Menge | 10 mg |
Synonym | cirazoline hydrochloride, 2-2-cyclopropylphenoxy methyl-4,5-dihydro-1h-imidazolium chloride, 2-2-cyclopropylphenoxy methyl-4,5-dihydro-1h-imidazole hydrochloride, cirazoline hcl, cirazoline hydrochloride, solid, 2-2-cyclopropylphenoxymethyl imidazoline hydrochloride, 1h-imidazole,2-2-cyclopropylphenoxy methyl-4,5-dihydro-,hydrochloride 1:1 |
InChI-Schlüssel | XFRXWLWUUDJHPB-UHFFFAOYSA-N |
SMILES | C1CC1C2=CC=CC=C2OCC3=NCCN3.Cl |
IUPAC-Name | 2-[(2-cyclopropylphenoxy)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
Molekulargewicht (g/mol) | 252.742 |
PubChem CID | 11957512 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'