Learn More
Tocris Bioscience™ CIA 1
COUP-TFII inhibitor
Marke: Tocris Bioscience™ 7494/10
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CIA 1 is an inhibitor of the nuclear receptor COUP-TFII (IC50 ranged from 1.2 μM to 7.6 μM, in prostate cancer cell lines). In vitro, CIA 1 inhibits growth of multiple prostate cancer cell lines and suppresses COUP-TFII activity to regulate its target genes. In vivo, CIA 1 reduces tumor growth in prostate cancer xenograft mouse models.Spezifikation
N-[2-(3,4-Dimethoxyphenyl)ethyl]-6-methylthieno[2,3-d]pyrimidin-4-amine hydrochloride | |
10 mg | |
C17H19N3O2S.HCl |
1049691-47-5 | |
Additional Nuclear Receptor Compounds | |
0.98 |