missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Säulen C18 ec Volumen: 1ml,Füllm.:100mg, Mat.:PP, mit PE-Filterelementen (VE=100St)

Brand:  Macherey-Nagel™ 730011

Product Code. 10014260

  • 203.00 € / 100 Stück
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Silika
SPE Hydra
45 μm
Säule für die Festphasenextraktion
Unpolar, hydrophob
60 μm
1 ml
HPLC-, CG-, TLC- und SPE-Anwendungen
Chromabond C 18 ec
Ja
Sphärisch
500 m2/g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Säulen C18 ec Volumen: 1ml,Füllm.:100mg, Mat.:PP, mit PE-Filterelementen (VE=100St) > Säulenkapazität: 1 ml; Adsorptionsgewicht: 100 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.