missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody.
Marke: Epredia™ MS-728-RQ
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.Host Species: Mouse
Clone: 56C6
Isotype: IgG1
Species Reactivity: Human and Rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD10 protein
Molecular Weight: 100kDa
Positive Control: Small intestine
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spezifikation
CD10 | |
Monoclonal | |
Unconjugated | |
Recombinant external domain of CD10 protein | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
56C6 | |
Mouse | |
12 mL | |
Oncogenesis | |
Ready-to-Use |