missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14388-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene is a member of the casein kinase I (CKI) gene family whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for this gene.Spezifikation
Casein Kinase 1 delta | |
Polyclonal | |
Unconjugated | |
CSNK1D | |
Casein Kinase 1 delta, casein kinase 1, delta, Casein kinase I isoform delta, CKI delta, CKId, CSNK1D, HCKID | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104318, 1453, 64462 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48730, Q06486, Q9DC28 | |
Csnk1d | |
Casein Kinase 1 delta Fusion Protein Ag5856 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |