missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Marke: Novus Biologicals NB600-103
478.80 EUR gültig bis 2025-12-31
BESTER Aktionspreis! Benutzen Sie den Promotions-Code "24272" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Calreticulin Polyclonal specifically detects Calreticulin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.
Spezifikation
| Calreticulin | |
| Polyclonal | |
| Western Blot 1:1000-1:10000, Simple Western 1:500, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:100. Use reported by customer review, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen reported in scientific literature (PMID 26643212), Knockout Validated, KnockDown Validated 1:2500 | |
| P14211 | |
| CALR | |
| Calreticulin Antibody was developed against a full-length protein to mouse Calreticulin [UniProt# P14211] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antisera |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
| Unconjugated | |
| Whole antisera with 0.1% Sodium Azide | |
| autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP | |
| Rabbit | |
| 48 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction | |
| 811 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction