missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-B Light Chain Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marke: R&D Systems AF5420-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BoNT-B Light Chain Polyclonal specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.
Spezifikation
| BoNT-B Light Chain | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BoNTB Light Chain, DcD | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6030102 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P10844 | |
| boNT/B | |
| E. coli-derived recombinant Clostridium BoNT-B Light Chain Pro2-His428 Accession # P10844 | |
| 25 μg | |
| Primary | |
| Detects C. botulinum BoNT-B Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant Light Chains of BoNT-A, -E, or recombinant Heavy Chains of BoNT-C, or -D is observed. | |
| C. botulinum | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur