Learn More
Brady™ BradyPrinter i7100 with Peel Function
The BradyPrinter i7100 industrial thermal transfer benchtop printer is ideal for reliable high volume and accurate printing on sleeves, tags, labels, tapes, cable markers and more.
Marke:  Brady™ 322880
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30242845
  • 2552.00 € / 1 Set
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
  • Print on the widest range of materials with confidence, including difficult to print materials
  • Peel configuration automatically peels labels from the liner
  • Prints ultra-sharp on labels as small as 4 mm with high accuracy
  • Designed to print thousands of labels per day, everyday
  • Easy-to-use touch screen interface to adjust printer settings quickly and easily
  • Ships with a licence for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.
Spezifikation
Spezifikation
Printer Kit
Color LCD
Direct Thermal, Thermal Transfer
9.9 in.
251.46 mm
289.56 mm
Monocolour
Brady Workstation Suite, Peel Function
256 MB RAM, 50 MB IFFS Flash, SD card up to 512 GB
150.00 mm/sec.
Brady Workstation, Windows
Windows™ 7, Windows™ 8, Windows™ 10, Windows™ Server 2012, Windows™ Server 2012 R2, Windows™ Server 2016
10 kg
2 years
100/240 V
i7100-600P-EU-BWS
Blue, Gray
Plastic
18.1 in.
459.74 mm
11.4 in.
459.74 x 251.46 x 289.56 mm
50.80 x 91.44 mm
English, Spanish, German, French, Dutch
i7100-600P-EU-BWS
600 dpi
Ethernet, I/O connection, Bluetooth (optional), Wi-Fi (optional), Serial, USB
22.05 lb.
CE, FCC class A, FCC class CB, CCC, UL
AC Adaptor
None
EU
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Zubehör anzeigen