missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Arrestin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10171-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.Spezifikation
Beta Arrestin 2 | |
Polyclonal | |
Unconjugated | |
ARRB2 | |
ARB2, ARR2, ARRB2, Arrestin beta 2, arrestin, beta 2, BARR2, Beta arrestin 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216869, 409 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32121, Q91YI4 | |
ARRB2 | |
Beta Arrestin 2 Fusion Protein Ag0227 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |