missing translation for 'onlineSavingsMsg'
Learn More

, Thermo Scientific™

Artikelnummer. 17322511 Alle ansehen Thermo Scientific Chemicals Produkte
Click to view available options
Menge:
100 g
1 kg
5 kg
Packungsgröße:
1 Kilogramm
100 Gramm
5 Kilogramm
This item is not returnable. View return policy

Product Code. 17322511

Brand: Thermo Scientific Acros 471231000

Bulk oder benutzerdefiniertes Format anfordern

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70131-50-9

Specifications

Chemischer Name oder Material Bentonit
Azidität 0.03 mg/KOH/g (Free Acidity)
Infrarotspektrum Entspricht der Struktur
Verpackung Kunststoffflasche
Menge 100 g
Titrierbare Säure ≤ 0.1 mg/KOH/g (Free Acidity)
Physikalische Form Granulate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.