Learn More
B7-H7 Monoclonal Antibody (MA57YW), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
144.00 € - 378.00 €
Spezifikation
| Antigen | B7-H7 |
|---|---|
| Klon | MA57YW |
| Konzentration | 5 μL/Test |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16300724
|
Invitrogen
17-6537-41 |
25 Tests | |||||||
16310724
![]() |
Invitrogen™
17-6537-42 |
100 Tests |
378.00 €
100 Stück |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
Description: This MA57YW monoclonal antibody recognizes human B7-H7 on live and paraformaldehyde-fixed cells. Applications Reported: This MA57YW antibody has been reported for use in flow cytometric analysis. Applications Tested: This MA57YW antibody has been pre-diluted and tested by flow cytometric analysis of the HDLM2 cell line. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.Spezifikation
| B7-H7 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q9UM44 | |
| HHLA2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MA57YW | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| 11148 | |
| B7-H7 recombinant protein | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| HHLA2 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.