Learn More
3M™ Aura™ Particulate Respirator
Particulate respirator with 3-panel design allows for greater facial movement
375.10 € - 718.30 €
Spezifikation
Produkttyp | Partikel-Atemschutzmaske |
---|---|
Mit Ausatemventil | Ohne Ventil |
Beschreibung
- Original 3-panel design allows for greater facial movement
- Low-resistance filter technology helps you breathe more easily
- Sculpted upper panel conforms to the nose and contours of the face for an effective seal
- Embossed top panel helps reduce eyewear fogging
- Chin tab designed for ease of positioning and adjustment
- Easy to store away with flat-fold design and individual wrapping
- Colour-coded headbands for easy identification: yellow for FFP1 (NFP 4)
- Sculpted nose panel helps conform to the nose and contours of the face and helps to improve compatibility with 3M eyewear
- Maximum usage level: Up to 4 x TLV
- 3M™ Cool Flow™ Valve helps reduce heat build-up in hot and humid conditions
- Available in large or small pack sizes
Spezifikation
Partikel-Atemschutzmaske | |
Ohne Ventil |