missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-TRAF2, MAP3K1 Protein Protein Interaction Antibody Pair, Abnova™
Marke: Abnova DI0490.1Set
Beinhaltet: TRAF2 rabbit purified polyclonal antibody (20μg); MAP3K1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Beschreibung
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TRAF2 protein, and the other against the MAP3K1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spezifikation
TRAF2 | |
Monoclonal/Polyclonal | |
TRAF2 | |
TRAF2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. TRAF2 rabbit purified polyclonal antibody (20 ug) 2. MAP3K1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
MGC:45012/TRAP/TRAP3 | |
Mouse, Rabbit | |
RUO | |
7186/4214 | |
Human | |
TRAF2 & MAP3K1 Protein Protein Interaction Antibody Pair |