missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ephrin B1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 343500
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. The signaling pathway downstream of the receptor is referred to as forward signaling while the signaling pathway downstream of the ephrin ligand is referred to as reverse signaling. Binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4. Together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration. EPHB4-mediated forward signaling controls cellular repulsion and segregation from EFNB2-expressing cells. May play a role in constraining the orientation of longitudinally projecting axons.
Spezifikation
| Ephrin B1 | |
| Polyclonal | |
| Unconjugated | |
| P98172, P52795 | |
| EFNB1 | |
| Synthetic peptide derived from the internal sequence of the mouse ephrin-B1 protein | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| EFNB1 | |
| EFL6, EFNB3, EPH related receptor transmembrane ligand ELK L3, EPH related receptor tyrosine kinase ligand 8, Ephrin B3, EPLG8, LERK8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13641, 1947 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur