Learn More
Ephrin A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 343300
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ephrin A1 Polyclonal antibody specifically detects Ephrin A1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Ephrin A1 | |
Polyclonal | |
Unconjugated | |
EFNA1 | |
AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13636, 1942 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P20827, P52793 | |
EFNA1 | |
Synthetic peptide derived from the C-terminal end of mouse ephrin-A1 protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |