Learn More
BrdU (Bromodeoxyuridine) Mouse, FITC, Clone: B44, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 347583
Beschreibung
Anti-BrdU, clone B44, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with spleen cells from BALB/c mice immunized with iodouridine-conjugated ovalbumin.
Bromodeoxyuridine (BrdU) is a uridine derivative that can be incorporated into DNA in place of thymidine. Anti-BrdU identifies BrdU (but not thymidine) in single-stranded DNA, free BrdU, or BrdU coupled to a protein carrier. The antibody also reacts with iodouridine.
Spezifikation
| BrdU (Bromodeoxyuridine) | |
| Monoclonal | |
| 25μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Iodouridine | |
| RUO | |
| Primary | |
| Purified |
| Flow Cytometry | |
| B44 | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. | |
| IgG1 κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.