missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-APC, CTNNB1 Protein Protein Interaction Antibody Pair, Abnova™
Marke: Abnova DI0250.1Set
Beinhaltet: APC rabbit purified polyclonal antibody (20μg); CTNNB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Beschreibung
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the APC protein, and the other against the CTNNB1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spezifikation
APC | |
Monoclonal/Polyclonal | |
APC | |
APC | |
1 Set | |
Primary | |
Antibody pair set content: 1. APC rabbit purified polyclonal antibody (20 ug) 2. CTNNB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
BTPS2/DP2/DP2.5/DP3/GS | |
Mouse, Rabbit | |
RUO | |
324/1499 | |
Human | |
APC & CTNNB1 Protein Protein Interaction Antibody Pair |