Learn More
Alpha 1B-Glycoprotein Mouse anti-Human, CoraLite 488, Clone: 4D4E5, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech CL488-66260-100UL
Weitere Details : Netto-Gewicht : 0.00010kg
Artikelnummer. 17204706
  • 367.00 € / 100 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Beschreibung
Beschreibung
Alpha-1B-glycoprotein (A1BG) is a plasma protein and is a member of the immunoglobulin superfamily. A1BG contains 474 amino acids and has 5 intrachain disulfide bonds. It has been reported that cysteine-rich secretory protein 3 (CRISP-3) is a specific and high-affinity ligand of A1BG. A1BG-CRISP-3 complex displays a similar function in protecting the circulation from a potentially harmful effect of free CRISP3 (PMID: 15461460).
Spezifikation
Spezifikation
Alpha 1B-Glycoprotein
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG1
Immunofluorescence
4D4E5
CoraLite 488
100 μL
1