Learn More
Ibidi 8 Well Chamber, Removable
Artikelnummer. 17256213
Encompass_Preferred
Click to view available options
:
Well Chamber
Artikelnummer. 17256213
Marke: Ibidi 80841
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
8 Well Chamber, Removable is a chamber slide with a removable 8 well silicone chamber.
  • Chamber slide with a removable 8 well silicone chamber for cell culture and immunofluorescence staining
  • for use with upright and inverted microscopes
  • bottom material: microscopy glass
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8
Sterilität Steril