missing translation for 'onlineSavingsMsg'
Learn More

[4-(tert-Butyl)phenyl]Methanesulfonylchlorid, Tech., Thermo Scientific™

101.00 € - 253.00 €

Chemische Identifikatoren

CAS 519056-61-2
Summenformel C11H15ClO2S
Molekulargewicht (g/mol) 246.749
MDL-Nummer MFCD04115413
InChI-Schlüssel NEJDUPGGBSPJLG-UHFFFAOYSA-N
Synonym 4-tert-butylphenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonylchloride, 4-tert-butylphenyl methanesulphonyl chloride, 4-1-butyl phenylmethanesulfonyl chloride, 4-tert-butyl phenyl methyl chlorosulfone
PubChem CID 2794645
IUPAC-Name (4-tert-Butylphenyl)methansulfonylchlorid
SMILES CC(C)(C)C1=CC=C(C=C1)CS(=O)(=O)Cl
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Product Code Brand Menge Price Quantity & Availability  
10300603
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
MO00924CB
250 mg
101.00 €
250 Milligramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
10664073
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
MO00924DA
1 g
253.00 €
1 Gramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

Specifications

Chemical Identifiers

519056-61-2
246.749
NEJDUPGGBSPJLG-UHFFFAOYSA-N
2794645
CC(C)(C)C1=CC=C(C=C1)CS(=O)(=O)Cl
C11H15ClO2S
MFCD04115413
4-tert-butylphenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonylchloride, 4-tert-butylphenyl methanesulphonyl chloride, 4-1-butyl phenylmethanesulfonyl chloride, 4-tert-butyl phenyl methyl chlorosulfone
(4-tert-Butylphenyl)methansulfonylchlorid

Specifications

[4-(tert-Butyl)phenyl]methanesulfonyl chloride
Bernsteinfarbene Glasflasche
MFCD04115413
NEJDUPGGBSPJLG-UHFFFAOYSA-N
(4-tert-Butylphenyl)methansulfonylchlorid
2794645
Tech.
519056-61-2
C11H15ClO2S
4-tert-butylphenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonyl chloride, 4-tert-butyl phenyl methanesulfonylchloride, 4-tert-butylphenyl methanesulphonyl chloride, 4-1-butyl phenylmethanesulfonyl chloride, 4-tert-butyl phenyl methyl chlorosulfone
CC(C)(C)C1=CC=C(C=C1)CS(=O)(=O)Cl
246.749
246.76
Technisch
Videos
Safety and Handling

Safety and Handling

GHS-H-Hinweis:
Verursacht schwere Augenschäden.
Verursacht schwere Verätzungen der Haut und schwere Augenschäden.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS-Signalwort:Gefahr

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[4-(tert-Butyl)phenyl]Methanesulfonylchlorid, Tech., Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.