Learn More
4-(2-Oxo-1-pyrrolidinyl)-benzaldehyd, 97 %, Thermo Scientific™
Chemische Identifikatoren
CAS | 36151-45-8 |
---|---|
Summenformel | C11H11NO2 |
Molekulargewicht (g/mol) | 189.214 |
MDL-Nummer | MFCD00454011 |
InChI-Schlüssel | VADZUJOCSAESJS-UHFFFAOYSA-N |
Synonym | 4-2-oxopyrrolidin-1-yl benzaldehyde, 4-2-oxo-1-pyrrolidinyl benzaldehyde, 4-2-oxo-pyrrolidin-1-yl-benzaldehyde, 4-2-oxopyrrolidinyl benzaldehyde, benzaldehyde,4-2-oxo-1-pyrrolidinyl, 4-2-oxo-pyrrolidin-1-yl benzaldehyde, 4-2-oxidanylidenepyrrolidin-1-yl benzaldehyde |
PubChem CID | 2794706 |
IUPAC-Name | 4-(2-Oxopyrrolidin-1-yl)benzaldehyd |
SMILES | C1CC(=O)N(C1)C2=CC=C(C=C2)C=O |
Produktcode | Marke | Menge | Verpackung | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Verpackung | Preis | Menge & Verfügbarkeit | ||||
10472793
|
Thermo Scientific Maybridge
MO01193CB |
250 mg | Bernsteinfarbene Glasflasche |
N/A
|
N/A | ||||
10288522
|
Thermo Scientific Maybridge
MO01193DA |
1 g | Bernsteinfarbene Glasflasche |
N/A
|
N/A | ||||
10672875
|
Thermo Scientific Maybridge
MO01193EA |
10 g | Bernsteinfarbene Glasflasche |
N/A
|
N/A | ||||
Beschreibung
Chemische Identifikatoren
36151-45-8 | |
189.214 | |
VADZUJOCSAESJS-UHFFFAOYSA-N | |
2794706 | |
C1CC(=O)N(C1)C2=CC=C(C=C2)C=O |
C11H11NO2 | |
MFCD00454011 | |
4-2-oxopyrrolidin-1-yl benzaldehyde, 4-2-oxo-1-pyrrolidinyl benzaldehyde, 4-2-oxo-pyrrolidin-1-yl-benzaldehyde, 4-2-oxopyrrolidinyl benzaldehyde, benzaldehyde,4-2-oxo-1-pyrrolidinyl, 4-2-oxo-pyrrolidin-1-yl benzaldehyde, 4-2-oxidanylidenepyrrolidin-1-yl benzaldehyde | |
4-(2-Oxopyrrolidin-1-yl)benzaldehyd |
Spezifikation
4-(2-Oxo-1-pyrrolidinyl)benzaldehyde | |
97.0 | |
C11H11NO2 | |
4-2-oxopyrrolidin-1-yl benzaldehyde, 4-2-oxo-1-pyrrolidinyl benzaldehyde, 4-2-oxo-pyrrolidin-1-yl-benzaldehyde, 4-2-oxopyrrolidinyl benzaldehyde, benzaldehyde,4-2-oxo-1-pyrrolidinyl, 4-2-oxo-pyrrolidin-1-yl benzaldehyde, 4-2-oxidanylidenepyrrolidin-1-yl benzaldehyde | |
C1CC(=O)N(C1)C2=CC=C(C=C2)C=O | |
189.214 | |
189.21 |
36151-45-8 | |
97% | |
MFCD00454011 | |
VADZUJOCSAESJS-UHFFFAOYSA-N | |
4-(2-Oxopyrrolidin-1-yl)benzaldehyd | |
2794706 | |
97% |
Sicherheit und Handhabung
GHS-H-Hinweis:
Verursacht Hautreizungen.
Verursacht schwere Augenreizung.
Kann die Atemwege reizen.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.