missing translation for 'onlineSavingsMsg'
Learn More

Chem-Lab 25LT Sodium hydroxide 1 mol/l (40 g NaOH / l H2O= 1 N (±0.002/20°C)), std sol volumetric analysis

Marke:  Chem-Lab CL05.1420.9025


25LT Sodium hydroxide 1 mol/l (40 g NaOH / l H2O= 1 N (±0.002/20°C)), std sol volumetric analysis

Artikelnummer. 11933233

  • ONLINE EXCLUSIVE PRICE
    317.00 €
    237.57 € / 25 Liter
    Sparen 79.43 €
    25% Rabatt

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Natronlauge 1 mol/l

Signal Word
  • Gefahr

Hazard Category
  • Hautreizende/-ätzende Wirkung Kategorie 1A
  • Stoffe und Gemische\, die gegenüber Metallen korrosiv sind Kategorie 1

Hazard Statement
  • H314-Verursacht schwere Verätzungen der Haut und schwere Augenschäden.
  • H290-Kann gegenüber Metallen korrosiv sein.

Precautionary Statement
  • P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P301+P330+P331-BEI VERSCHLUCKEN: Mund ausspülen. KEIN Erbrechen herbeiführen.
  • P308+P311-BEI Exposition oder falls betroffen: GIFTINFORMATIONSZENTRUM/ Arzt/ anrufen.
  • P305+P351+P338-BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.

Supplemental information
  • MIXTURE LIST-Enthält: Sodium hydroxide, pellets a.r.

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.