missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-ThioUTP tetrasodium salt, Tocris Bioscience™
Click to view available options
Menge:
1 mg
Beschreibung
Potent and selective P2Y2 agonist (EC50 values are 0.035, 0.35 and 1.5 μM for hP2Y2, hP2Y4 and hP2Y6 receptors respectively).
Spezifikation
Spezifikation
Chemischer Name oder Material | 2-ThioUTP tetrasodium salt |
CAS | 1343364-70-4 |
Summenformel | C9H15N2Na4O14P3S |
Menge | 1 mg |
Synonym | 2-thioutp tetrasodium salt |
InChI-Schlüssel | ZMGCYKHWZHBMKG-ODQFIEKDSA-N |
SMILES | C1=CN(C(=S)NC1=O)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na].[Na] |
IUPAC-Name | [[(2R,3S,4R,5R)-3,4-dihydroxy-5-(4-oxo-2-sulfanylidenepyrimidin-1-yl)oxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium |
Molekulargewicht (g/mol) | 592.159 |
PubChem CID | 131846931 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'