missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pyridylethylamine dihydrochloride, Tocris Bioscience™
Click to view available options
Menge:
50 mg
Beschreibung
Histamine H1 receptor agonist that produces vasoconstriction in vivo.
Spezifikation
Spezifikation
Chemischer Name oder Material | 2-Pyridylethylamine dihydrochloride |
CAS | 3343-39-3 |
Summenformel | C7H12Cl2N2 |
Menge | 50 mg |
Synonym | 2-pyridylethylamine dihydrochloride, pyridine, 2-2-aminoethyl-, dihydrochloride, 2-2-aminoethyl pyridine dihydrochloride, c7h10n2.2hcl, 2-pyridineethanamine dihydrochloride, 2-pyridin-2-ylethanamine dihydrochloride, 2-pyridin-2-yl ethanamine dihydrochloride |
InChI-Schlüssel | GPIFKSYYILNJNR-UHFFFAOYSA-N |
SMILES | C1=CC=NC(=C1)CCN.Cl.Cl |
IUPAC-Name | 2-pyridin-2-ylethanamine;dihydrochloride |
Molekulargewicht (g/mol) | 195.087 |
PubChem CID | 201148 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'