Learn More
DiscoverX 2-(methylthio)-ADP
Artikelnummer. 17362499
Encompass
Click to view available options
:
5 mg
Artikelnummer. 17362499
Marke: DiscoverX 921177
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen
2-(Methylthio)adenosine 5′-diphosphate (2-methylthio-ADP) induces platelet aggregation and inhibits cAMP accumulation in platelets in-vitro. It's 3-5 times more active than ADP as an aggregating agent, and 150-200 times more active as a cAMP accumulation inhibitor.
2-(Methylthio)adenosine 5′-diphosphate (2-methylthio-ADP) induces platelet aggregation and inhibits cAMP accumulation in platelets in-vitro. It's 3-5 times more active than ADP as an aggregating agent, and 150-200 times more active as a cAMP accumulation inhibitor. Potent agonist of the P2Y1, P2Y12, and P2Y13 purinoceptors. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer.
Spezifikation
Menge 5 mg