missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ 1A-116
Rac1 inhibitor; blocks Rac1-P-Rex1 interaction
Marke: Tocris™ 6701/50
848.36 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
1A-116 is a Rac1 inhibitor. Reduces intracellular Rac1-GTP levels and blocks Rac1-P-Rex1 interaction in vitro. Exhibits no effect on GTPase activity of closely related Cdc42. Displays antiproliferative, anti-invasive and pro-apoptotic activity in several cancer cell lines. Exhibits antimetastatic activity in vivo. Also restores antiproliferative effects of Tamoxifen (Cat. No. 6342) in resistant breast cancer cell.Spezifikation
N-(3,5-Dimethylphenyl)-N'-[2-(trifluoromethyl)phenyl]guanidine | |
50 mg | |
307.31 | |
98% |
1430208-73-3 | |
C16H16F3N3 | |
Hemmstoff |