missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIIA/CD16a Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Marke: R&D Systems FAB107512V-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Fc gamma RIIIA/CD16a Monoclonal antibody specifically detects Fc gamma RIIIA/CD16a in Human samples. It is validated for Flow Cytometry
Spezifikation
| Fc gamma RIIIA/CD16a | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGR3A, FCGRIII, FcgRIIIA, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA | |
| Mouse myeloma cell line NS0-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2829B | |
| Flow Cytometry | |
| P08637 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 2214 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur