missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody (5C2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Marke: Novus Biologicals NBP1-04321
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Spezifikation
| alpha-Synuclein | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Mouse | |
| 14.46 kDa | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5C2 | |
| Unconjugated | |
| P37840 | |
| SNCA | |
| This alpha-Synuclein Antibody (5C2) was developed against recombinant human alpha-synuclein (61-95) purified from E. coli (NP_0003360). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human alpha-Synuclein (61-95 aa) mAb, clone 5C2, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human alpha-synuclein protein. This specifically recognizes alpha-synuclein only, particularly the NAC domain of alpha-synuclein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur