Isobutyrylchlorid, 98 %, Thermo Scientific Chemicals
Allgemein
32.20 EUR gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Weitere Details:
Weitere Details: CAS: 79-30-1 Netto-Gewicht: 0.10000kg Transport: UN number : 2395 Chem class : 3 Pack group : II
Menge 100 g
Impressum: RUO – Research Use Only
Artikelnummer. 11495937
Menge Preis
1 32.36  € / 100 Gramm
Auf Lager, versandfertig: 4
Zum Warenkorb hinzufügen
  • Beschreibung und Spezifikation
  • Alternative Produkte
  • Informationsmaterial
  • Sicherheit und Handhabung
Beschreibung und Spezifikation
Chemische Identifikatoren
CAS 79-30-1
Summenformel C4H7ClO
Molekulargewicht (g/mol) 106.549
MDL-Nummer MFCD00000717
InChI-Schlüssel DGMOBVGABMBZSB-UHFFFAOYSA-N
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
PubChem CID 62325
IUPAC-Name 2-Methylpropanoylchlorid
SMILES CC(C)C(=O)Cl
Spezifikation
Chemischer Name oder Material Isobutyryl chloride
CAS 79-30-1
Schmelzpunkt ∽-90°C
Dichte 1.017
Siedepunkt 91°C to 93°C
Flammpunkt 8°C (46°F)
Prozentgehaltsbereich 0.98
Summenformel C4H7ClO
Brechungsindex 1.407
MDL-Nummer MFCD00000717
Menge 100 g
UN-Nummer UN2395
Beilstein 605393
Empfindlichkeit Moisture sensitive
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Synonym isobutyryl chloride,2-methylpropionyl chloride,propanoyl chloride, 2-methyl,isobutanoyl chloride,isobutyric acid chloride,isobutyroyl chloride,isobutyrylchloride,dimethylacetyl chloride,chloro isopropyl ketone,alpha-methylpropionyl chloride
Löslichkeitsinformationen Miscible with chloroform,glacial acetic acid,ether,toluene,methylene chloride and benzene.
InChI-Schlüssel DGMOBVGABMBZSB-UHFFFAOYSA-N
SMILES CC(C)C(=O)Cl
IUPAC-Name 2-Methylpropanoylchlorid
Molekulargewicht (g/mol) 106.549
PubChem CID 62325
Formelmasse 106.55
Reinheit (%) 98%
Physikalische Form Flüssigkeit
Anwendungen
Isobutylchlorid wird bei der Herstellung von vollständig mit Isobutyramid terminierten hyperverzweigten Polyethyleniminen verwendet. Es spielt eine wichtige Rolle bei der Trennung und Identifizierung von Shikonin und seinen Esterderivaten in der Wurzelepidermis von Echium Italicum L. Es fungiert als Akylierungsmittel für die Ketiminderivate von alfa-Amino-Estern, Nukleosiden und Pyrrolen. Darüber hinaus wird es bei der Herstellung organischer Peroxide verwendet. Außerdem wird es als Zwischenprodukt für Farbstoffe, Textilhilfsmittel und Peroxidverbindungen verwendet.
Löslichkeit
Mischbar mit Chloroform, Eisessigsäure, Äther, Toluol, Methylenchlorid und Benzol.
Hinweise
Feuchtigkeitsempfindlich. Nicht kompatibel mit Basen, Wasser, starken Oxidationsmitteln und Alkoholen.
Alternative Produkte
Alternative Verpackung
Katalognummer Marke Beschreibung Bestand Preis
Menge
11405947 Thermo Scientific Alfa Aesar ISOBUTYRYL CHLORIDE, 98% 500G 1 95.17 €
Zum Warenkorb hinzufügen
Alternative
Katalognummer Marke Beschreibung Bestand Preis
Menge
10255730 Thermo Scientific Acros Isobutyryl chloride, 98%, 250ml 3 62.51 €
Zum Warenkorb hinzufügen
Informationsmaterial
Technische Unterlagen
Sicherheit und Handhabung
[Isobutyryl chloride]
[Gefahr]
[{code=ACUTE TOX. 2, sequence=1, description=Akute Toxizität Kategorie 2}, {code=ACUTE TOX. 4, sequence=2, description=Akute Toxizität Kategorie 4}, {code=EYE DAM. 1, sequence=3, description=Schwere Augenschädigung/Augenreizung Kategorie 1}, {code=FLAM. LIQ. 2, sequence=4, description=Entzündbare Flüssigkeit Kategorie 2}, {code=MET. CORR. 1, sequence=5, description=Stoffe und Gemische\, die gegenüber Metallen korrosiv sind Kategorie 1}, {code=SKIN CORR. 1A, sequence=6, description=Hautreizende/-ätzende Wirkung Kategorie 1A}]
[{code=H225, sequence=1, description=Flüssigkeit und Dampf leicht entzündbar.}, {code=H290, sequence=2, description=Kann gegenüber Metallen korrosiv sein.}, {code=H302, sequence=3, description=Gesundheitsschädlich bei Verschlucken.}, {code=H314, sequence=4, description=Verursacht schwere Verätzungen der Haut und schwere Augenschäden.}, {code=H318, sequence=5, description=Verursacht schwere Augenschäden.}, {code=H330, sequence=6, description=Lebensgefahr bei Einatmen.}]
[{code=P210, sequence=51, description=Von Hitze, heißen Oberflächen, Funken, offenen Flammen und anderen ndquellen fernhalten. Nicht rauchen.}, {code=P260, sequence=52, description=Staub/Rauch/Gas/Nebel/Dampf/Aerosol nicht einatmen.}, {code=P280, sequence=53, description=Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.}, {code=P301+P330+P331, sequence=54, description=BEI VERSCHLUCKEN: Mund ausspülen. KEIN Erbrechen herbeiführen.}, {code=P305+P351+P338, sequence=55, description=BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.}, {code=P310, sequence=56, description=Sofort GIFTINFORMATIONSZENTRUM/Arzt/anrufen.}]
[{code=MIXTURE LIST, sequence=1, description=Enthält: Isobutyrylchloride}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS05, sequence=2, description=CORROSION}, {code=GHS06, sequence=3, description=SKULL & CROSSBONES}]
Produktidentifikator
  • Isobutyryl chloride
Signalwort
  • Gefahr
Gefahrenkategorie
  • Akute Toxizität Kategorie 2
  • Akute Toxizität Kategorie 4
  • Schwere Augenschädigung/Augenreizung Kategorie 1
  • Entzündbare Flüssigkeit Kategorie 2
  • Stoffe und Gemische\, die gegenüber Metallen korrosiv sind Kategorie 1
  • Hautreizende/-ätzende Wirkung Kategorie 1A
Gefahrenhinweis
  • H225- Flüssigkeit und Dampf leicht entzündbar.
  • H290- Kann gegenüber Metallen korrosiv sein.
  • H302- Gesundheitsschädlich bei Verschlucken.
  • H314- Verursacht schwere Verätzungen der Haut und schwere Augenschäden.
  • H318- Verursacht schwere Augenschäden.
  • H330- Lebensgefahr bei Einatmen.
Sicherheitshinweis
  • P210- Von Hitze, heißen Oberflächen, Funken, offenen Flammen und anderen ndquellen fernhalten. Nicht rauchen.
  • P260- Staub/Rauch/Gas/Nebel/Dampf/Aerosol nicht einatmen.
  • P280- Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
  • P301+P330+P331- BEI VERSCHLUCKEN: Mund ausspülen. KEIN Erbrechen herbeiführen.
  • P305+P351+P338- BEI KONTAKT MIT DEN AUGEN: Einige Minuten lang behutsam mit Wasser spülen. Vorhandene Kontaktlinsen nach Möglichkeit entfernen. Weiter spülen.
  • P310- Sofort GIFTINFORMATIONSZENTRUM/Arzt/anrufen.
Ergänzende Informationen
  • MIXTURE LIST- Enthält: Isobutyrylchloride