CAS RN 1071466-61-9
CAS RN 1071466-61-9
IUPAC Name:
4-{2-[4-(acetyloxy)-3-(prop-2-en-1-yl)phenyl]propan-2-yl}-2-(prop-2-en-1-yl)phenyl acetate
Synonyme:
4-{2-[4-(acetyloxy)-3-(prop-2-en-1-yl)phenyl]propan-2-yl}-2-(prop-2-en-1-yl)phenyl acetate
Molekulargewicht (g/mol):
392.5
Summenformel:
C25H28O4
InChi Key:
GJTZQXNRNNDGRG-UHFFFAOYSA-N
SMILES:
CC(=O)OC1=C(CC=C)C=C(C=C1)C(C)(C)C1=CC(CC=C)=C(OC(C)=O)C=C1
1
–
1
von
1
Ergebnisse
1
3,3'-Diallylbisphenol A-Diacetat, Epoxidharz, EC-392, Thermo Scientific Chemicals
CAS: 1071466-61-9 Summenformel: C25H28O4 Molekulargewicht (g/mol): 392.495 MDL-Nummer: MFCD22200539 InChI-Schlüssel: GJTZQXNRNNDGRG-UHFFFAOYSA-N Synonym: 2,2'-diallyl bisphenol a diacetate ether,4,4'-isopropylidene bis 2-allylphenol acetate,phenyl ester epoxy curative hybrid of bisphenol a,3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392,4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate PubChem CID: 59215472 IUPAC-Name: [4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetat SMILES: CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C