CAS RN 1071466-53-9
CAS RN 1071466-53-9
IUPAC Name:
4-{[4-(acetyloxy)phenyl]methyl}phenyl propanoate
Synonyme:
4-{[4-(acetyloxy)phenyl]methyl}phenyl propanoate
Molekulargewicht (g/mol):
298.34
Summenformel:
C18H18O4
InChi Key:
JARUUDOWTQBKKU-UHFFFAOYSA-N
SMILES:
CCC(=O)OC1=CC=C(CC2=CC=C(OC(C)=O)C=C2)C=C1
1
–
1
von
1
Ergebnisse
1
Bisphenol F-Acetatpropionat, Epoxidharz, EC-298, Thermo Scientific Chemicals
CAS: 1071466-53-9 Summenformel: C18H18O4 Molekulargewicht (g/mol): 298.338 MDL-Nummer: MFCD22380705 InChI-Schlüssel: JARUUDOWTQBKKU-UHFFFAOYSA-N Synonym: 4-4-acetyloxy phenyl methyl phenyl propanoate,bisphenol f acetate propionate, epoxy curative, ec-298 PubChem CID: 59317436 IUPAC-Name: [4-[(4-acetyloxyphenyl)methyl]phenyl]-Popanoat SMILES: CCC(=O)OC1=CC=C(C=C1)CC2=CC=C(C=C2)OC(=O)C