CAS RN 107-12-0
CAS RN 107-12-0
IUPAC Name:
propanenitrile
Synonyme:
propionitrile
Molekulargewicht (g/mol):
55.08
Summenformel:
C3H5N
InChi Key:
FVSKHRXBFJPNKK-UHFFFAOYSA-N
SMILES:
CCC#N
1
–
3
von
3
Ergebnisse
1
Propionitril, 99 %, extra rein, Thermo Scientific Chemicals
CAS: 107-12-0 Summenformel: C3H5N Molekulargewicht (g/mol): 55.08 MDL-Nummer: MFCD00001948 InChI-Schlüssel: FVSKHRXBFJPNKK-UHFFFAOYSA-N Synonym: propionitrile,ethyl cyanide,propiononitrile,cyanoethane,propylnitrile,propionic nitrile,ether cyanatus,hydrocyanic ether,ethylkyanid,propannitril PubChem CID: 7854 ChEBI: CHEBI:26307 IUPAC-Name: Propanenitril SMILES: CCC#N
2
Propionitril, 99 %, Thermo Scientific Chemicals
CAS: 107-12-0 Summenformel: C3H5N Molekulargewicht (g/mol): 55.08 MDL-Nummer: MFCD00001948 InChI-Schlüssel: FVSKHRXBFJPNKK-UHFFFAOYSA-N Synonym: propionitrile,ethyl cyanide,propiononitrile,cyanoethane,propylnitrile,propionic nitrile,ether cyanatus,hydrocyanic ether,ethylkyanid,propannitril PubChem CID: 7854 ChEBI: CHEBI:26307 IUPAC-Name: Propanenitril SMILES: CCC#N