CAS RN 1067-87-4
CAS RN 1067-87-4
IUPAC Name:
diethyl (prop-2-en-1-yl)phosphonate
Synonyme:
diethyl prop-2-en-1-ylphosphonate
Molekulargewicht (g/mol):
178.17
Summenformel:
C7H15O3P
InChi Key:
YPJHXRAHMUKXAE-UHFFFAOYSA-N
SMILES:
CCOP(=O)(CC=C)OCC
1
–
1
von
1
Ergebnisse
1
Diethylallylphosphonat, 97 %, Thermo Scientific Chemicals
CAS: 1067-87-4 Summenformel: C7H15O3P Molekulargewicht (g/mol): 178.17 MDL-Nummer: MFCD00015134 InChI-Schlüssel: YPJHXRAHMUKXAE-UHFFFAOYSA-N Synonym: diethyl allylphosphonate,diethyl prop-2-en-1-ylphosphonate,diethyl-allylphosphonate,acmc-1bprn,phosphonic acid, 2-propenyl-, diethyl ester,allylphosphonic acid diethyl,3-diethoxyphosphoryl-prop-1-ene,allylphosphonic acid diethyl ester,allyl phosphonic acid diethyl ester PubChem CID: 573014 IUPAC-Name: 3-Diethoxyphosphorylprop-1-en SMILES: CCOP(=O)(CC=C)OCC