CAS RN 1055301-07-9
CAS RN 1055301-07-9
IUPAC Name:
1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine dihydrochloride
Synonyme:
1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine dihydrochloride
Molekulargewicht (g/mol):
465.12
Summenformel:
C14H8Cl2F10N2
InChi Key:
JXHUHXWIKZGKIE-UHFFFAOYNA-N
SMILES:
Cl.Cl.NC(C(N)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
1
–
1
von
1
Ergebnisse
1
(R,R)-(+)-1,2-Bis(2,3,4,5,6-Pentafluorphenyl)-1,2-Ethandiamindihydrochlorid (Forts. 10-20 Gew.% Mono-HCl), 95 %, ee 99 %, Thermo Scientific™
CAS: 1055301-07-9 Summenformel: C14H8Cl2F10N2 Molekulargewicht (g/mol): 465.12 MDL-Nummer: MFCD09265309,MFCD09953690 InChI-Schlüssel: JXHUHXWIKZGKIE-UHFFFAOYNA-N Synonym: 1r,2r-1,2-bis 2,3,4,5,6-pentafluorophenyl ethane-1,2-diamine dihydrochloride,r,r-1,2-bis 2,3,4,5-pentafluorophenyl-1,2-ethanediamine dihydrochloride, ee,r,r-+-1,2-bis 2,3,4,5,6-pentafluorophenyl-1,2-ethanediamine dihydrochloride cont. 10-20wt% mono hcl PubChem CID: 73995081 IUPAC-Name: (1R,2R)-1,2-bis(2,3,4,5,6-pentafluorophenyl)ethan-1,2-diamin;Dihydrochlorid SMILES: Cl.Cl.NC(C(N)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F