CAS RN 104-40-5
CAS RN 104-40-5
IUPAC Name:
4-nonylphenol
Synonyme:
4-nonylphenol N-nonylphenol nonylphenol (isomer mixture) P-nonylphenol
Molekulargewicht (g/mol):
220.36
Summenformel:
C15H24O
InChi Key:
IGFHQQFPSIBGKE-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC1=CC=C(O)C=C1
1
–
3
von
3
Ergebnisse
1
4-n-Nonylphenol, 98+ %, Thermo Scientific Chemicals
CAS: 104-40-5 Summenformel: C15H24O Molekulargewicht (g/mol): 220.356 MDL-Nummer: MFCD00002396 InChI-Schlüssel: IGFHQQFPSIBGKE-UHFFFAOYSA-N Synonym: 4-n-nonylphenol,p-nonylphenol,phenol, 4-nonyl,para-nonylphenol,p-n-nonylphenol,phenol, nonyl,phenol, p-nonyl,nonylphenol mixed,phenol, nonyl derivs.,4-nonyl phenol PubChem CID: 1752 ChEBI: CHEBI:34440 IUPAC-Name: 4-Nonylphenol SMILES: CCCCCCCCCC1=CC=C(C=C1)O