CAS RN 10326-28-0
CAS RN 10326-28-0
IUPAC Name:
cadmium(2+) hexahydrate diperchlorate
Synonyme:
cadmium(2+) hexahydrate diperchlorate
Molekulargewicht (g/mol):
419.4
Summenformel:
CdCl2H12O14
InChi Key:
PQJZOTJHMRJOEM-UHFFFAOYSA-L
SMILES:
O.O.O.O.O.O.[Cd++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
1
–
1
von
1
Ergebnisse
1
Cadmiumperchlorat-Hexahydrat, weiß, kristallin, Thermo Scientific Chemicals
CAS: 10326-28-0 Summenformel: CdCl2H12O14 Molekulargewicht (g/mol): 419.396 MDL-Nummer: MFCD00015992 InChI-Schlüssel: PQJZOTJHMRJOEM-UHFFFAOYSA-L Synonym: cadmium perchlorate hexahydrate,acmc-1c6hb,cd.2clo4.6h2o,cadmium ii perchlorate hexahydrate PubChem CID: 71317310 IUPAC-Name: Cadmium(2+);diperchlorat;hexahydrat SMILES: O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]