CAS RN 10326-27-9
CAS RN 10326-27-9
Bariumchlorid-Dihydrat, zertifiziertes AR für Analysen, Fisher Chemical
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L IUPAC-Name: Barium(2+)-Dihydrat-Dichlorid SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid Dihydrat, 99+ %, Thermo Scientific Chemicals
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 SMILES: O.O.[Cl-].[Cl-].[Ba++]
Sonderaktionen verfügbar
Bariumchlorid-Dihydrat, 99+ %, zur Analyse, Thermo Scientific Chemicals
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC-Name: Barium(2+); Dichlorid; Dihydrat SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, ACS, Thermo Scientific Chemicals
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, 99+ %, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC-Name: Barium(2+); Dichlorid; Dihydrat SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, Puratronic, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC-Name: Barium(2+); Dichlorid; Dihydrat SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, Puriss, p.a. plus, ACS-Reagenz, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: Bariumchlorid-Dihydrat,Barium2+-Dihydrat-Dichlorid,Barium2+-Dichlorid-Dihydrat,Bariumchlorid, ACS,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC-Name: Barium(2+); Dichlorid; Dihydrat SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, reinst, SLR, Fisher Chemical
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L IUPAC-Name: Barium(2+)-Dihydrat-Dichlorid SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid-Dihydrat, ≥99 %, Honeywell™ Fluka™
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: Bariumchlorid-Dihydrat,Barium2+-Dihydrat-Dichlorid,Barium2+-Dichlorid-Dihydrat,Bariumchlorid, ACS,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 SMILES: O.O.[Cl-].[Cl-].[Ba++]
Bariumchlorid Dihydrat, ACS-Reagenz, ≥99 %, Honeywell™ Fluka™
CAS: 10326-27-9 Summenformel: BaCl2H4O2 Molekulargewicht (g/mol): 244.26 MDL-Nummer: MFCD00149154 InChI-Schlüssel: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: Bariumchlorid-Dihydrat,Barium2+-Dihydrat-Dichlorid,Barium2+-Dichlorid-Dihydrat,Bariumchlorid, ACS,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC-Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]