CAS RN 103-73-1
CAS RN 103-73-1
IUPAC Name:
ethoxybenzene
Synonyme:
phenyl ethyl alcohol
Molekulargewicht (g/mol):
122.17
Summenformel:
C8H10O
InChi Key:
DLRJIFUOBPOJNS-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1
1
–
2
von
2
Ergebnisse
1
Phenetol, 98+ %, Thermo Scientific Chemicals
CAS: 103-73-1 Summenformel: C8H10O Molekulargewicht (g/mol): 122.167 MDL-Nummer: MFCD00009090 InChI-Schlüssel: DLRJIFUOBPOJNS-UHFFFAOYSA-N Synonym: phenetole,benzene, ethoxy,ethyl phenyl ether,phenetol,phenyl ethyl ether,phenoxyethane,a phenoxyethane,ether, ethyl phenyl,unii-rb8lu2c57f PubChem CID: 7674 ChEBI: CHEBI:67129 IUPAC-Name: Ethoxybenzol SMILES: CCOC1=CC=CC=C1
2
Phenetol, 99 %, Thermo Scientific Chemicals
CAS: 103-73-1 Summenformel: C8H10O Molekulargewicht (g/mol): 122.17 MDL-Nummer: MFCD00009090 InChI-Schlüssel: DLRJIFUOBPOJNS-UHFFFAOYSA-N Synonym: phenetole,benzene, ethoxy,ethyl phenyl ether,phenetol,phenyl ethyl ether,phenoxyethane,a phenoxyethane,ether, ethyl phenyl,unii-rb8lu2c57f PubChem CID: 7674 ChEBI: CHEBI:67129 IUPAC-Name: Ethoxybenzol SMILES: CCOC1=CC=CC=C1