CAS RN 1023595-17-6
CAS RN 1023595-17-6
IUPAC Name:
(1H-indazol-4-yl)boronic acid
Synonyme:
1H-indazol-4-ylboronic acid
Molekulargewicht (g/mol):
161.96
Summenformel:
C7H7BN2O2
InChi Key:
BGZZJZIZRARGGZ-UHFFFAOYSA-N
SMILES:
OB(O)C1=C2C=NNC2=CC=C1