CAS RN 10108-64-2
CAS RN 10108-64-2
Calciumchlorid, 99 %, wasserfrei, Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, ACS-Reagenz, wasserfrei, Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, wasserfrei, ACS, 99.0 % min., Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, ultratrocken, 99.998 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, weißes Pulver, 99.99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, ultratrocken, 99.996 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, 98 %, Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, 99.99 %, (Metallspurenanalyse), Thermo Scientific Chemicals
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: cadmium chloride,cadmium dichloride,caddy,vi-cad,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]
Cadmiumchlorid, Honeywell
CAS: 10108-64-2 Summenformel: CdCl2 Molekulargewicht (g/mol): 183.31 MDL-Nummer: MFCD00010916 InChI-Schlüssel: YKYOUMDCQGMQQO-UHFFFAOYSA-L Synonym: Cadmiumchlorid,Cadmiumdichlorid,Caswell Nr. 135,Cadmium-II-chlorid,kadmiumchlorid,caswell no. 135,ccris 114,hsdb 278,cadmium ii chloride PubChem CID: 24947 ChEBI: CHEBI:35456 IUPAC-Name: Dichlorcadmium SMILES: [Cl-].[Cl-].[Cd++]