CAS RN 1008415-02-8
CAS RN 1008415-02-8
IUPAC Name:
(3-chloro-4-cyanophenyl)boronic acid
Synonyme:
3-chloro-4-cyanophenylboronic acid
Molekulargewicht (g/mol):
181.38
Summenformel:
C7H5BClNO2
InChi Key:
QCJTUOIMDNJEHR-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(Cl)=C(C=C1)C#N
1
–
1
von
1
Ergebnisse
1
3-Chlor-4-Cyanobenzolboronsäure, 96%, Thermo Scientific Chemicals
CAS: 1008415-02-8 Summenformel: C7H5BClNO2 Molekulargewicht (g/mol): 181.382 InChI-Schlüssel: QCJTUOIMDNJEHR-UHFFFAOYSA-N Synonym: 3-chloro-4-cyanophenyl boronic acid,boronic acid, b-3-chloro-4-cyanophenyl,acmc-2097sr,3-chloro-4-cyanophenyl boronicacid PubChem CID: 46738932 IUPAC-Name: (3-Chlor-4-cyanphenyl)boronsäure SMILES: B(C1=CC(=C(C=C1)C#N)Cl)(O)O