CAS RN 100-39-0
CAS RN 100-39-0
IUPAC Name:
(bromomethyl)benzene
Synonyme:
benzyl bromide
Molekulargewicht (g/mol):
171.04
Summenformel:
C7H7Br
InChi Key:
AGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILES:
BrCC1=CC=CC=C1
1
–
2
von
2
Ergebnisse
1
Benzylbromid 98 %, Thermo Scientific Chemicals
CAS: 100-39-0 MDL-Nummer: MFCD00000172 InChI-Schlüssel: AGEZXYOZHKGVCM-UHFFFAOYSA-N Synonym: benzyl bromide,bromomethyl benzene,alpha-bromotoluene,bromophenylmethane,benzene, bromomethyl,phenylmethyl bromide,1-bromotoluene,benzylbromide,cyclite PubChem CID: 7498 ChEBI: CHEBI:59858 IUPAC-Name: brommethylbenzol SMILES: C1=CC=C(C=C1)CBr
2
Benzylbromid, 99 %, Thermo Scientific Chemicals
CAS: 100-39-0 Summenformel: C7H7Br Molekulargewicht (g/mol): 171.037 MDL-Nummer: MFCD00000172 InChI-Schlüssel: AGEZXYOZHKGVCM-UHFFFAOYSA-N Synonym: benzyl bromide,bromomethyl benzene,alpha-bromotoluene,bromophenylmethane,benzene, bromomethyl,phenylmethyl bromide,1-bromotoluene,benzylbromide,cyclite PubChem CID: 7498 ChEBI: CHEBI:59858 IUPAC-Name: brommethylbenzol SMILES: C1=CC=C(C=C1)CBr