missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSPC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA5100676
Informations supplémentaires : Netto-Gewicht : 0.01000kg
Description
Antibody detects endogenous levels of total PSPC1.
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.Spécification
PSPC1 | |
Polyclonal | |
Unconjugated | |
PSPC1 | |
5730470C09Rik; AI327109; AI449052; mPSP1; Paraspeckle component 1; paraspeckle protein 1; PSP1; Pspc1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55269, 66645 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8R326, Q8WXF1 | |
PSPC1 | |
A synthesized peptide derived from human PSPC1(Accession Q8WXF1), corresponding to amino acid residues Q489-Y523. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |